Learn More
Medchemexpress LLC 3-O-Acetylhamaudol | 30358-88-4 | 10 MG

Supplier: Medchemexpress LLC HYN689210MG
3-O-Acetylhamaudol is a chemical compound isolated from Angelica polymorpha. It has been observed to demonstrate anti-tumor activity through anti-angiogenesis and activation of intestinal intraepithelial lymphocytes.
- Molecular formula: C17H18O6
- Molecular weight: 318.32
- Physical form: Solid
- Color: White to off-white
- Isolated from Angelica polymorpha
- Demonstrates anti-tumor activity
- Supports anti-angiogenesis
- Activates intestinal intraepithelial lymphocytes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.