Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-O-Acetylhamaudol | 30358-88-4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN68925MG

Encompass_Preferred

3'-O-Acetylhamaudol is a compound isolated from Angelica japonica roots. It exhibits anti-tumor activity through dual actions: anti-angiogenesis and intestinal intraepithelial lymphocyte activation.

  • Isolated from *Angelica japonica* roots
  • Exhibits anti-tumor activity
  • Supports anti-angiogenesis and intestinal intraepithelial lymphocyte activation
  • Purity greater than 98%
  • Type of compound: Flavonoid
  • Molecular formula: C17H18O6
  • Molecular weight: 318.325
  • Intended for reference standard and R&D use

Catalog No. 50-004-18100


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.