Learn More
Medchemexpress LLC 3-O-acetyltylosin-d3 | 98.0% | 961.16 g/mol | C48H76D3NO18 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYW585906S500UG
Encompass_Preferred
3-O-Acetyltylosin-d3 is a deuterium-labeled analogue of 3-O-acetyltylosin intended for use as an internal standard or tracer in analytical and drug-development studies. It provides a stable isotopic reference for quantitative assays; lot-specific purity and storage conditions are available in the certificate of analysis.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Molecular formula C48H76D3NO18; molecular weight 961.16 g/mol.
  • Supplied as a 500 μg package suitable for analytical workflows.
  • High nominal purity (98%) for consistent assay performance.
  • Certificate of Analysis provides lot-specific purity and storage instructions.
  • Suitable for mass spectrometry and pharmacokinetic tracer studies.
Catalog No. 50-002-84444