Learn More
Apexbio Technology LLC 3'-O-Methyl-CTP 10ul (100 mM)
SDP
Supplier:  Apexbio Technology LLC B80370.01
Encompass_Preferred
3p-O-Methyl-CTP is a modified nucleoside triphosphate It features a methyl group at the 3 -hydroxyl position of the ribose enabling enzymatic incorporation into nucleic acids and modulating physicochemical properties such as RNA stability and protein interactions 3p-O-Methyl-CTP exerts its biological activity by serving as a substrate for RNA synthesis introducing sugar modifications that impact RNA-protein recognition mechanisms and gene expression regulation Based on these characteristics 3p-O-Methyl-CTP holds research potential in molecular biology and epigenetics facilitating studies on RNA modifications nucleic acid modulation and the functional analysis of epigenetic processes
Catalog No. 50-000-88143