Learn More
Medchemexpress LLC 3-O-Methyldopa (standard) | 300-48-1 | 99.9% | 211.22 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY113468AR10MG
Encompass_Preferred
3-O-Methyldopa (Standard) is the analytical standard of 3-O-Methyldopa, intended for research and analytical applications. It is a metabolite of L-DOPA, formed by catechol-O-methyltransferase (COMT), and competitively inhibits the pharmacodynamics of l-DOPA and dopamine. The compound is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard
  • Suitable for HPLC, GC and MS applications
  • Metabolite of L-DOPA
  • Competitively inhibits l-DOPA and dopamine pharmacodynamics
Catalog No. 50-004-37904