Learn More
Medchemexpress LLC 3'-O-methylguanosine | 10300-27-3 | 100.0% | 297.27 | C11H15N5O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW091784100MG
Encompass_Preferred
3'-O-methylguanosine is a methylated nucleoside analog used as a research reagent and analytical standard. It functions as an RNA chain terminator and has been reported to inhibit early vaccinia virus-specific RNA synthesis and plaque formation in vitro.
  • Functions as an RNA chain terminator
  • Reported to inhibit vaccinia virus early RNA synthesis in vitro
  • Available as solid and solution formats, including 100 MG solid and 10 mM in DMSO solution
  • High purity suitable for analytical applications
  • Certificate of analysis and safety data sheet available
Catalog No. 50-002-09439