Learn More
Selleck Chemical LLC 3-O-Methylquercetin E6044-1MG
SDP
Supplier:  Selleck Chemical LLC E60441MG
Encompass_Preferred
3-O-Methylquercetin is a flavone isolated from Rhamnus nakaharai Hayata (Rhamnaceae) an inhibitor of cAMPand cGMP-phosphodiesterases(PDE) with an IC50 of 13 8 M and 14 3 M respectively It also inhibits -secretase with an IC50 of 6 5 M It exhibits antiviral activity and has been used in folk medicine for constipation inflammation tumors and asthma
Catalog No. 50-000-41073