Learn More
TARGETMOL CHEMICALS INC 3-O-p-Coumaroylquinic acid 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN70481MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 3-O-p-Coumaroylquinic acid, hydroxycinnamic acid derivative, is isolated from a 70% methanolic Crataegus extract (Crataegi folium cum flore). Purity 99.81%
Catalog No. 50-288-0253