Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Oxetanone | 6704-31-0 | 99.5% | C3H4O2 | 10 G
SDP

Supplier:  Medchemexpress LLC HY7644010G

Encompass_Preferred

3-Oxetanone is a drug intermediate used to construct spirocyclic compounds with anti-glioblastoma properties.

  • Purity: 99.54%
  • Appearance: Colorless to light yellow liquid
  • Molecular weight: 72.06 g/mol
  • Boiling point: 140 °C
  • Flash point: 26.1 °C

Catalog No. 50-004-33857


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.