Learn More
AdipoGen 3-oxo-C6-L-HSL
SDP
Supplier:  AdipoGen CDXO0057M050
Encompass_Preferred
Chemical. CAS 143537-62-6. Formula C10H15NO4. MW 213.2. Synthetic. N-3-Oxohexanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. At increased populations of bacteria, localized higher concentrations of 3OC6-HSL, an endogenous ligand to transcriptional factor LuxR, leads to increased production of both the AHL synthase and proteins responsible for bioluminescence. Numerous other species of bacteria also employ 3OC6-HSL in cell-to-cell communication. An autoinducer of P. fischeri luciferase and a specfic genetic regulator that is unrelated to at least one of the enzyme systems that it induces.
Catalog No. 50-256-0310