Learn More
Medchemexpress LLC 3-Oxopentanedioic acid | 542-05-2 | 146.10 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW007752100G
Encompass_Preferred
3-Oxopentanedioic acid is a dicarboxylic acid that is easily brominated and serves as an intermediate in organic chemical synthesis. It is also an aliphatic β-dicarbonyl acid, useful as a model compound for studying the formation mechanism of brominated trihalomethanes during chlorination, bromination, and ozonation of saline water.
  • Easily brominated dicarboxylic acid
  • Intermediate in organic chemical synthesis
  • Aliphatic β-dicarbonyl acid
  • Model compound for studying brominated trihalomethanes formation
Catalog No. 50-004-46442