Learn More
Medchemexpress LLC 3-Oxopentanedioic acid | 542-05-2 | 146.10 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW007752500G
Encompass_Preferred
3-Oxopentanedioic acid is a dicarboxylic acid that is readily brominated and serves as an intermediate in organic chemical synthesis. It is also an aliphatic β-dicarbonyl acid that can be utilized as a model compound to study the formation mechanism of brominated trihalomethanes during chlorination, bromination, and ozonation of saline water.
  • Can be used as an intermediate in organic chemical synthesis.
  • Functions as a model compound for relevant structures in natural organic matter for environmental studies.
  • Available with high purity.
  • Soluble in H₂O (100 mg/mL) and DMSO (100 mg/mL).
Catalog No. 50-004-48678