Learn More
Medchemexpress LLC 3-Penten-2-one | 625-33-2 | 99.7% | 84.12 | 10 G

Supplier: Medchemexpress LLC HY3474810G
3-Penten-2-one (Ethylideneacetone) inhibits nitric oxide production and inducible nitric oxide synthase expression via heme oxygenase-1 induction in RAW264.7 macrophages activated with Lipopolysaccharides (LPS). This chemical is for research use only and not intended for human or animal therapeutic use.
- Inhibits nitric oxide production
- Inhibits inducible nitric oxide synthase expression
- Functions as a biochemical assay reagent
- Identified as an endogenous metabolite
- Long-term storage stability at -20°C for pure form
- Solvent storage at -80°C for 6 months
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.