Learn More
Medchemexpress LLC 3-phenoxypropyl bromide | 588-63-6 | MFCD00000256 | >95.0% | 215.09 g/mol | C9H11BrO | 100 G
SDP
Supplier:  Medchemexpress LLC HYW019627100G
Encompass_Preferred
3-Phenoxypropyl bromide (CAS 588-63-6) is an organic biochemical reagent used as an alkylating agent and building block in organic and medicinal chemistry. Supplied as a liquid, it is employed in synthesis to introduce a 3-phenoxypropyl group in target molecules and is used for intermediate and reagent applications in research laboratories.
  • Used as an alkylating reagent in organic synthesis.
  • Suitable as a building block and intermediate for medicinal chemistry.
  • High purity (typically >95% by GC) for research use.
  • Liquid form allows straightforward handling and reaction setup.
  • Available in multiple pack sizes to support small- and large-scale work.
Catalog No. 50-002-99545