Learn More
Medchemexpress LLC 3-phenoxypropyl bromide | 588-63-6 | MFCD00000256 | 97.1% | 215.09 g/mol | C9H11BrO | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01962710G
Encompass_Preferred
3-Phenoxypropyl bromide (CAS 588-63-6) is an organic alkyl bromide used as a reagent and intermediate in organic synthesis and life-science research. It is supplied for laboratory research use only and is commonly employed to introduce a 3-phenoxypropyl moiety in etherification and nucleophilic substitution reactions.
  • Versatile reagent for etherification and substitution reactions.
  • Serves as a building block in small-molecule synthesis.
  • Offered in laboratory-scale packaging (10 g-100 g) for research workflows.
  • For research use only; not intended for human or clinical use.
  • CAS number 588-63-6 enables unambiguous chemical identification.
Catalog No. 50-003-00970