Learn More
Medchemexpress LLC HY-125759 5mg Medchemexpress, 3-Phenyltoxoflavin CAS:32502-63-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1257595MG
Encompass_Preferred
Medchemexpress, HY-125759 5mg 3-Phenyltoxoflavin CAS:32502-63-9 3-Phenyltoxoflavin, a derivative of Toxoflavin, is an Hsp90 inhibitor, with a Kd of 585 nM for the interaction of Hsp90-TPR2A. 3-Phenyltoxoflavin has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1083