Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 35UM3 P
Supplier:  Gilson Company Inc GAA78
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 333; Sieve opening; 35um
Catalog No. 50-871-422