Learn More
Medchemexpress LLC 3-pyridinecarboxylic acid, 6-[5-(ethoxycarbonyl)-2-thiazolyl]-, ethyl ester | 1821370-70-0 | 99.9% | 306.34 | C14H14N2O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10306810MM 1ML
Encompass_Preferred
Diethyl-pythiDC is a small-molecule inhibitor of collagen prolyl 4-hydroxylases (CP4Hs) used in research to modulate collagen hydroxylation. It inhibits CP4H activity in cultured cells at concentrations that do not cause iron deficiency and shows minimal cytotoxicity in tested cell lines.
  • High purity (99.9%) suitable for research applications.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities (5 mg, 10 mg, 25 mg, 50 mg, 100 mg).
  • Stable under recommended storage: powder at -20 °C (long term) or 4 °C; in solvent at -80 °C or -20 °C.
  • Molecular weight 306.34 and chemical formula C14H14N2O4S.
  • Effective in cell-based assays without affecting iron-regulated proteins at tested concentrations.
Catalog No. 50-002-46275