Learn More
Medchemexpress LLC 3-pyridinecarboxylic acid, 6-[5-(ethoxycarbonyl)-2-thiazolyl]-, ethyl ester | 1821370-70-0 | MFCD31715412 | 99.9% | 306.34 g/mol | C14H14N2O4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10306850MG
Encompass_Preferred
Diethyl-pythiDC is a small-molecule inhibitor of collagen prolyl 4-hydroxylases (CP4Hs) supplied for research use. It is provided as a high-purity research chemical, available as a solid or as a ready-made 10 mM solution in DMSO, and is used in cellular and in vitro studies to probe CP4H activity and collagen biosynthesis.
  • Inhibits collagen prolyl 4-hydroxylases (CP4Hs) for research applications.
  • High purity, 99.9% as reported by the manufacturer.
  • Molecular weight 306.34 g/mol and formula C14H14N2O4S.
  • Available as solid or 10 mM solution in DMSO for convenience.
  • Suitable for in vitro and cellular assays to study collagen synthesis.
Catalog No. 50-002-46148