Learn More
Medchemexpress LLC 3-Pyridinemethanol | 100-55-0 | 99.58% | 100 G

Supplier: Medchemexpress LLC HYB0893100G
3-Pyridinemethanol, also known as Nicotinyl alcohol, is a pyridine derivative. It functions as a cholesterol-lowering agent and can be selectively oxidized to vitamin B3.
- Functions as a cholesterol-lowering agent
- Can be selectively oxidized to vitamin B3
- Target: CETP
- Pathway: Metabolic enzyme/protease
- Storage (pure form): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
- Appearance: Light yellow to light brown liquid
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.