Learn More
Gilson Company Inc 3"SIEVE BR/SS FULL #100
Supplier:  Gilson Company Inc V3CF #100
Encompass_Preferred
3" Sieve, Brass/Stainless, Full Height, No. 1003in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 100 (150µm) openings and full height brass frame. The overall height is 1-3/4in and the stacked height is 1-1/8in.
Catalog No. NC2147393