Learn More
ATCC 3 strain; Tagged Genomic DNA; Even Mix
Supplier: ATCC MSA1014/DEL
This NGS Standard is a mock microbial community that can be used as a spike-in control for 16S rRNA and shotgun metagenomic sequencing assays. This product comprises genomic DNA prepared from three fully characterized and authenticated ATCC Genuine Cultures that contain unique synthetic 16S tags incorporated into the bacterial genome. Each tag consists of four artificial variable regions (corresponding to V1 through V4 of 16S rRNA gene) flanked by conserved regions for PCR amplification. Total cells: 6.0 x 107 genome copies/vial ± 1 log* *Whole cell concentrations indicate ATCC manufacturing specifications and are provided as a reference only