Learn More
Medchemexpress LLC 3-sulfopropanoic acid | 44826-45-1 | MFCD19347139 | 95.0% | 154.14 g/mol | C3H6O5S | 5 MG

Supplier: Medchemexpress LLC HYW2517795MG
3-Sulfopropanoic acid is an endogenous sulfonic acid used as a research reagent in biochemical and metabolic studies. Supplied as a solid, it is suitable for analytical and preparative laboratory applications.
- Purity 95.0%.
- Molecular formula C3H6O5S.
- Molecular weight 154.14 g/mol.
- Appearance: colorless to off-white solid.
- Available in small milligram quantities for research use.
- Contains a propanoic acid backbone with a sulfonic acid functional group, enabling solubility in polar solvents.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.