Learn More
Medchemexpress LLC 3'-TBDMS-Bz-rA phosphoramidite | 129451-75-8 | 98.38% | 988.19 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY2170310MM/1ML
Encompass_Preferred
3'-TBDMS-Bz-rA Phosphoramidite is a phosphoramidite monomer used in the synthesis of oligonucleotides. Oligonucleotides are short-chain nucleic acids, composed of deoxyribonucleotides or ribonucleotides, that can specifically "silence" genes by complementary base pairing with DNA, mRNA, or pre-mRNA, thereby inhibiting protein expression.
  • Used in the synthesis of oligonucleotides.
  • Enables specific gene silencing through complementary base pairing.
  • Inhibits the expression of numerous proteins.
Catalog No. 50-004-03208