Learn More
U.S. Pharmacopeia 3-tert-Butyl-4-hydroxyanisole, 121-00-6, MFCD00040484, 200mg
Supplier:  U.S. Pharmacopeia 1083100
Encompass
Molecular Formula C11H16O2, Molecular Weight 180.245, Synonyms: 2-tert-Butyl-4-methoxyphenol * 3-BHA, 2-(1,1-dimethylethyl)-4-methoxy-Phenol
Catalog No. NC9664985