Learn More
Hogentogler & Co Inc 3" Test Sieve, No. 25 Mesh, Half Height, Stainless Frame - Stainless Cloth
SDP
Supplier:  Hogentogler & Co Inc 1712
Encompass_Preferred
Model 1712 is a 3" test sieve with a No. 25 mesh. This sieve is constructed with a Stainless Frame - Stainless Cloth and is Half Height. Proudly made in the USA.
Catalog No. NC1545385