Learn More
Cayman Chemical 3-ThIatetradecnoIc AcId 10mg
SDP
Supplier:  Cayman Chemical 9050010 MG
Encompass_Preferred
3-thia TDA is an analog of the 14-carbon saturated fatty acid myristic acid, wherein the C-3 carbon has been replaced by sulfur in a thioether linkage. When chronically administered to rats, 3-thia TDA acts as a peroxisome inducer (PPAR ligand), increases fatty acid oxidation, and lowers plasma lipid levels.{3828} Incorporation of 3-thia TDA into dietary regimens at 150 and 300 mg/kg in rats induces the formation of oleic acid via .DELTA.9-desaturase, and dose-dependently increases .DELTA.9-desaturase mRNA levels.{4942}
Catalog No. 50-289-6077