Learn More
Bellco Glass, Inc. 3 Tier Rocker Tray
SDP
Supplier:  Bellco Glass, Inc. 774000030
Encompass_Preferred
Tray is 11" long, 7" deep and 4 5/8" high.There is approximately 2" between the trays.It's sst and there are no liners.. MADE IN THE USA
Catalog No. 50-194-6102