Learn More
Medchemexpress LLC 3-Tolylboronic acid | 17933-03-8 | 135.96 | 25 G

Supplier: Medchemexpress LLC 25GHYW007381
3-Tolylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is intended for research use only.
- Can be used as a biological material
- Can be used as an organic compound for life science research
- Utilized in Suzuki coupling reactions
- Stable as powder for 3 years at -20°C
- Stable as powder for 2 years at 4°C
- Stable in solvent for 6 months at -80°C
- Stable in solvent for 1 month at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.