Learn More
Medchemexpress LLC 3-Trifluoromethyl-5,6,7,8-tetrahydro-triazolo[4,3-a]pyrazine hydrochloride | 762240-92-6 | MFCD09817638 | 98.0% | C6H8ClF3N4 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY76120
Encompass_Preferred
3-Trifluoromethyl-5,6,7,8-tetrahydro-triazolo[4,3-a]pyrazine hydrochloride is a biochemical reagent primarily used as an intermediate for sitagliptin phosphate. It is also suitable for life science-related research as a biological material or organic compound. This product appears as a white to off-white solid.
  • Biochemical reagent
  • Intermediate for sitagliptin phosphate
  • Utilized as a biological material
  • Used as an organic compound for life science research
  • Appears as a white to off-white solid
  • For research use only
Catalog No. 50-003-58966