Learn More
Medchemexpress LLC HY-108331 5mg Medchemexpress, 3-TYP CAS:120241-79-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1083315MG
Encompass_Preferred
Medchemexpress, HY-108331 5mg 3-TYP CAS:120241-79-4 3-TYP is a selective SIRT3 inhibitor, with an IC50 of 16 nM, more potent over SIRT1 (IC50=88 nM), SIRT2 (IC50=92 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8532