Learn More
Cayman Chemical 3 Z 6 Z 9 Z 12 Z 15 ZOctadeca
SDP
Supplier:  Cayman Chemical 100097312.5 MG
Encompass_Preferred
A microalgal PUFA; has been used as a chemotaxonomic biomarker for microalgae and dinoflagellates
Catalog No. 50-289-0169