Learn More
Medchemexpress LLC 30-azido-4,7,10,13,16,19,22,25,28-nonaoxatriacontanoic acid | 1670249-37-2 | MFCD29938006 | >97.0% | 511.56 | C21H41N3O11 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY130475250MG
Encompass_Preferred
Azido-PEG9-acid is an azide-terminated polyethylene glycol (9-unit) carboxylic acid linker for bioconjugation and click chemistry. It provides an azide functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and a carboxylic acid handle for further coupling.
  • Azide functional group for click chemistry (CuAAC).
  • Nine ethylene glycol units provide hydrophilicity and spacer function.
  • Terminal carboxylic acid enables standard coupling reactions.
  • Suitable for use as an ADC or PROTAC linker and other bioconjugation applications.
  • Stable as a pure solid when stored at low temperature according to manufacturer guidance.
Catalog No. 50-002-04124