Learn More
Sentry Air Systems 30 in Ductless Spray Booth
SDP
Supplier:  Sentry Air Systems SS330DSH
Encompass
The 30 in. Wide Portable Spray Booth provides a respiratory engineering control for aerosol spray to help protect the operator from exposure. This negative-pressure enclosure helps workplaces meet OSHA and EPA regulations while providing safety controls for employees. This system can be used for a variety of aerosol spray applications including aerosol particle testing and spray epoxy. The 30 in. Portable Spray Booth utilizes a three-step filtration process: 1- Internal Spray Filter, 2- HEPA filter, and 3- Activated Carbon. This energy efficient and low maintenance system helps protect employees by removing hazardous aerosol spray particles and unwanted odor from the airflow. The 30 Portable Spray Booth features a ductless design, removing the need for external ducting and makeup air in turn reducing operating costs. Approximate Airflow = 50 350 CFM
Catalog No. NC2885523