Learn More
ELKO FILTERING CO 30 MICRON NYLON MESH
SDP
Supplier:  ELKO FILTERING CO 0330/18
Encompass
Nylon Monofilament Precision Woven Mesh - 30 Micron Opening, Open Area 18%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-30/18
Catalog No. NC0478162