Learn More
Electron Microscopy Sciences Pop-Up Slide Holder, 30 place, 1ea
SDP
Supplier:  Electron Microscopy Sciences 71522
Encompass_Preferred
Holds 1x3" slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed.
Catalog No. 50-931-159