Learn More
Teledyne ISCO, Inc 300 Ml Seal Kit, Organic For Stainless Steel Pumps. Requires Qty 2 For Dual Pumps
Supplier:  Teledyne ISCO, Inc 890212
Encompass
300 ml Seal Kit, Organic for stainless steel pumps. Requires qty 2 for dual pumps
Catalog No. 50-211-9683