Learn More
Chemglass Life Sciences Chemglass 300 μL Automation Clear Tips for Hamilton Systems, Sterile, Barcoded (96/pk, 2,304/cs)
SDP
Supplier:  Chemglass Life Sciences CLS3499A043
Encompass_Preferred
Chemglass 300 μL automation clear tips are sterile, barcoded pipette tips designed for automated liquid-handling workflows. Supplied in blister-packed nests (96 per pack, 2,304 per case), they support high-throughput applications including genomics, proteomics, immunoassays, metabolomics, and biopharmaceutical workflows.
  • 300 μL volume.
  • Clear tip for visual confirmation.
  • Sterile, blister-packed nests.
  • Barcoded for inventory and tracking.
  • 96 per pack, 2,304 per case.
  • Designed for automated, high-throughput pipetting.
  • Suitable for genomics, proteomics, immunoassays, and metabolomics.
Catalog No. 50-273-3268