Learn More
Chemglass Life Sciences 300 μL Automation filter tips for Hamilton systems, clear, sterile, 96/rack (CLS-3499A-064)
SDP
Supplier:  Chemglass Life Sciences CLS3499A064
Encompass_Preferred
Chemglass 300 μL automation filter tips are sterile, clear pipette tips supplied in nested racks with barcodes to streamline automated workflows. They are intended for high-throughput liquid handling applications and provide aerosol protection during sample transfer.
  • 300 μL nominal volume.
  • Clear polypropylene construction for visual inspection.
  • Filter included for aerosol and contamination protection.
  • Sterile and ready for immediate use.
  • 96 tips per rack; 10 racks per pack, 50 racks per case.
  • Rack barcode included for automated identification.
  • Nested racks in boxed packaging for easy handling.
Catalog No. 50-273-3257