Learn More
Chemglass Life Sciences Chemglass Automation Conductive Filter Tips for Hamilton Systems, 300 μL, Sterile, 96/Pack (Blister Pack, Nest)
SDP
Supplier:  Chemglass Life Sciences CLS3499A012
Encompass_Preferred
Automation conductive filter tips designed for use with Hamilton automated pipetting systems. These sterile, conductive filter tips have a 300 μL nominal volume and are supplied in blister-packed nests for high-throughput automation workflows. Each pack contains 96 tips; cases contain 2,304 tips, and packaging includes a barcode to support inventory and automation.
  • 300 μL nominal volume.
  • Sterile, single-use.
  • Conductive filter tip design.
  • Blister-packed nested format; 96 tips per pack, 2,304 tips per case.
  • Medical-grade polypropylene construction.
  • Packaging includes barcode for automation and inventory.
Catalog No. 50-273-3280