Learn More
Chemglass Life Sciences 300 μL Automation Conductive Filter Tips for Hamilton Automated Liquid Handlers, Sterile, Barcode, 96/Rack
SDP
Supplier:  Chemglass Life Sciences CLS3499A032
Encompass_Preferred
Sterile 300 μL conductive filter tips designed for automated liquid handling systems. Made from medical-grade conductive polypropylene, these aerosol-resistant tips include rack-level barcodes for automation and are DNase/RNase free, suitable for molecular biology and precision liquid handling.
  • Sterile, electron beam sterilized.
  • Conductive polypropylene for tip detection and grounding.
  • Aerosol-resistant filter reduces cross-contamination risk.
  • Rack-level barcode supports automated inventory and tracking.
  • Packaging: 96 tips per rack, 10 racks per pack, 50 racks per case.
  • DNase- and RNase-free for sensitive workflows.
  • Manufactured in a controlled clean-room environment.
Catalog No. 50-273-3271