Learn More
CELLTREAT Scientific Products 300cm2 Tissue Culture Flask - Vent Cap, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229361
Encompass_Preferred
CELLTREAT Scientific Products 300cm¿ Tissue Culture Treated Flask. Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Vent cap and maximum volume of 850mL. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 3/re-sealable bag, 18/case.
Catalog No. 50-403-871