Learn More
Chemglass Life Sciences Chemglass Pipette Tips 300 μL, Clear, Bulk Packed, Non-sterile (CLS-3499E-300)
SDP
Supplier:  Chemglass Life Sciences CLS3499E300
Encompass_Preferred
Chemglass 300 μL clear pipette tips are bulk-packed, non-sterile tips designed for routine laboratory liquid handling. They are molded from an ultra-clear polymer for good visibility and are manufactured to minimize common contaminants, making them suitable for general pipetting applications.
  • Made from ultra-clear polypropylene for improved visibility.
  • Nominal volume 300 μL suitable for standard pipettors.
  • Bulk-packed format for high-volume workflows.
  • Offered in 1,000 per pack and 10,000 per case quantities.
  • Non-sterile, intended for general laboratory use.
  • Manufactured without common lubricants or cadmium-containing pigments.
Catalog No. 50-273-3207