Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer Techne 3028900 Portable Dry Block Calibrator, 120VAC

Supplier:  Cole-Parmer 1610197

Encompass_Preferred

  • Wide temperature range
    Independent over temperature cutout
  • Maximum temperature of 430C/806F
  • Built rugged for portable field use
  • Fast heat up and cool down
  • 10 insert formats available
  • Temperature sensor burnout protection
  • Lightweight, only 9 lbs
  • Low cost

Catalog No. 50-196-1441


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.