Learn More
Medchemexpress LLC 306-012B | 2566523-06-4 | 99.9% | 1251.08 g/mol | C59H115N3O8S8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW59053210MG
Encompass_Preferred
306-O12B is an ionizable cationic lipid used to formulate lipid nanoparticles (LNPs) for mRNA delivery in research. It combines a high molecular weight and tail-branched structure with defined solubility and storage properties to support formulation development and in vivo testing.
  • Ionizable cationic lipid suitable for LNP formulation.
  • High purity (99.9%) for reproducible results.
  • Soluble in DMSO at 100 mg/mL for concentrated stock solutions.
  • Compatible with multiple in vivo formulation protocols.
  • Clear storage guidelines for long-term and solution stability.
  • Available in small research pack sizes for screening and optimization.
Catalog No. 50-002-84418