Learn More
Medchemexpress LLC 306-O12B | 2566523-06-4 | 99.9% | 1251.08 | C59H115N3O8S8 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW59053225MG
Encompass_Preferred
An ionizable cationic lipid used to formulate lipid nanoparticles (LNPs) for delivery of mRNA. The compound is a tail-branched, bioreducible lipidoid offered in liquid or powder forms for research use, with recommended storage conditions to preserve stability.
  • Ionizable cationic lipid suitable for LNP formulation.
  • Tail-branched, bioreducible lipidoid structure enabling efficient mRNA encapsulation.
  • High purity (99.9%) to support reproducible research results.
  • Offered as small research-scale packs and as DMSO solutions for direct use.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
Catalog No. 50-002-85007