Learn More
SAI Infusion Technologies Sterile Blunt Needles, 30G, 0.5"", 25/pack
SDP
Supplier:  SAI Infusion Technologies B3050
Encompass
This item has a minimum qty of 3 per supplier requirements.
30G0.5 Blunt Needles with plastic hub and stainless steel blunt needle. Blunt tip needles are useful for a variety of laboratory tasks, including connection to indwelling catheters. SAI's smooth, burr-free stainless steel needles prevent damage to the lumen of the catheter during dosing, sampling, flushing or locking. These needles are sterile and individually packaged, 25/pk. Non-sterile blunt needles also available. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply.  Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
Catalog No. NC9770101