Learn More
Electron Microscopy Sciences 30-Slide Staining Dish and Lid only, 3¾" x 3⅞" x 3-3/8" (H)
SDP
Supplier:  Electron Microscopy Sciences 71430DL
Encompass_Preferred
These glass staining dishes are made of a heavy glass with snug fit lids. Lids are included. Dishes have a gasket seal. Accomodates 30 slides.
I.D.: 3¾" x 3⅞" x 3-3/8" (H)
Dimensions: 4" x 4⅛" x 4" (H)
Catalog No. 50-192-9695