Learn More
Chemglass Life Sciences Chemglass 30 μL Automation Low-Retention Tips for Agilent Systems (CLS-3499A-601)
SDP
Supplier:  Chemglass Life Sciences CLS3499A601
Encompass_Preferred
Chemglass 30 μL automation low-retention clear tips (CLS-3499A-601) are sterile, medical-grade polypropylene pipette tips designed for Agilent automated liquid-handling systems. They reduce liquid retention, improve transfer accuracy with viscous samples, and support high-throughput workflows via 384-rack packaging.
  • Low-retention design reduces liquid hanging on tip walls and improves transfer accuracy.
  • Medical-grade polypropylene construction; DNase/RNase free.
  • Electron beam sterilized and produced in a clean-room environment.
  • Packaged 384 per rack, 10 racks per pack, 50 racks per case for automation workflows.
  • Suitable for high-throughput automated liquid handling and viscous liquids.
Catalog No. 50-273-3229