Learn More
Ace Glass, Inc. Vacuum manifold, 310mm long, (3) stopcock ports with 14/20inner ends
SDP
Supplier:  Ace Glass, Inc. 874510
Encompass_Preferred
310mm long, single tube vacuum manifold featuring 3 stopcock ports with 14/20 inner joints. Distance between ports is 95mm which allows for use of flasks up to 3L.
Catalog No. 50-976-310